Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LGALS3 SIRNA Assays ›
          Gene Symbol:
          Lgals3
          Gene Name:
          lectin, galactoside-binding, soluble, 3
          Gene Aliases:
          AGE-R3, CBP30, L-34, gal-3
          Chromosome Location:
          Chr.15: 24153602 - 24165537 on Build Rnor_6.0
          UniGene ID:
          Rn.764
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 54395

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lgals3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031832.1 NP_114020.1 5 632
          GenBank mRNA BC089054.1 - 5 620 7312891
          FQ214799.1 - 5 914
          FQ219929.1 - 5 632
          FQ219987.1 - 5 632
          FQ220157.1 - 5 632
          FQ220174.1 - 5 631
          FQ220215.1 - 5 634
          FQ220224.1 - 5 626
          FQ220313.1 - 5 632
          FQ220316.1 - 6 862
          FQ220348.1 - 5 632
          FQ220361.1 - 5 791
          FQ220369.1 - 5 632
          FQ220439.1 - 5 631
          FQ220450.1 - 5 632
          FQ220455.1 - 6 859
          FQ220487.1 - 6 832
          FQ220505.1 - 5 632
          FQ220541.1 - 5 632
          FQ220563.1 - 5 632
          FQ220590.1 - 5 634
          FQ220652.1 - 5 632
          FQ220701.1 - 5 631
          FQ220837.1 - 5 631
          FQ220917.1 - 5 632
          FQ220938.1 - 5 632
          FQ220946.1 - 5 632
          FQ220952.1 - 5 632
          FQ221377.1 - 5 632
          FQ223467.1 - 5 634
          FQ223525.1 - 6 424
          FQ223531.1 - 5 632
          FQ229211.1 - 5 636
          FQ229249.1 - 5 632
          FQ229259.1 - 5 632
          FQ229266.1 - 5 632
          FQ229277.1 - 5 629
          FQ229326.1 - 5 632
          FQ229348.1 - 5 632
          FQ229362.1 - 5 632
          FQ229490.1 - 5 632
          FQ229494.1 - 5 632
          FQ229511.1 - 5 632
          FQ229560.1 - 5 634
          FQ229562.1 - 5 634
          FQ229577.1 - 5 632
          FQ229661.1 - 5 632
          FQ229821.1 - 5 632
          FQ229967.1 - 5 634
          FQ229990.1 - 5 634
          FQ229991.1 - 5 634
          FQ229995.1 - 5 632
          FQ230027.1 - 5 632
          FQ230050.1 - 5 635
          FQ230072.1 - 5 632
          FQ230121.1 - 5 639
          FQ230149.1 - 5 634
          J02962.1 - 5 632
          KF639951.1 - 4 592
          KF639952.1 - 4 592
          KF639953.1 - 4 592
          KF639954.1 - 4 592
          KF639955.1 - 5 571
          KF639956.1 - 5 571
          KF639957.1 - 5 571
          KF639958.1 - 5 571
          M13697.1 - 2 220

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          monocyte chemotaxis
          mRNA processing
          signal transduction
          positive regulation of cell proliferation
          RNA splicing
          positive regulation of serotonin secretion
          extracellular matrix organization
          neutrophil chemotaxis
          epithelial cell differentiation
          regulation of T cell proliferation
          negative regulation of apoptotic process
          innate immune response
          positive regulation of angiogenesis
          negative regulation of endocytosis
          eosinophil chemotaxis
          macrophage chemotaxis
          negative regulation of T cell receptor signaling pathway
          positive chemotaxis
          regulation of T cell apoptotic process
          mononuclear cell migration
          positive regulation of mononuclear cell migration
          positive regulation of calcium ion import
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of cell proliferation in bone marrow
          negative regulation of immunological synapse formation
          negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          negative regulation of extrinsic apoptotic signaling pathway
          IgE binding
          Fc-gamma receptor I complex binding
          chemoattractant activity
          laminin binding
          poly(A) RNA binding
          monosaccharide binding
          disaccharide binding
          advanced glycation end-product receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0