Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PAX5 SIRNA Assays ›
          Gene Symbol:
          PAX5
          Gene Name:
          paired box 5
          Gene Aliases:
          ALL3, BSAP
          Chromosome Location:
          Chr.9: 36833274 - 37035949 on Build GRCh38
          UniGene ID:
          Hs.654464
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 5952

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PAX5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001280547.1 NP_001267476.1 2, 3 668
          NM_001280548.1 NP_001267477.1 2, 3 668
          NM_001280549.1 NP_001267478.1 2, 3 668
          NM_001280550.1 NP_001267479.1 2, 3 668
          NM_001280551.1 NP_001267480.1 1, 2 502
          NM_001280552.1 NP_001267481.1 2, 3 668
          NM_001280553.1 NP_001267482.1 2, 3 668
          NM_001280554.1 NP_001267483.1 2, 3 668
          NM_001280556.1 NP_001267485.1 1, 2 502
          NM_016734.2 NP_057953.1 2, 3 668
          NR_103999.1 2, 3 668
          NR_104000.1 2, 3 668
          XM_005251481.4 XP_005251538.1 2, 3 1289
          XM_011517896.2 XP_011516198.1 2, 3 2138
          XM_011517897.2 XP_011516199.1 2, 3 1293
          GenBank mRNA AF080573.1 - 1, 2 38
          AY463952.1 - 2, 3 230
          AY463953.1 - 2, 3 230
          AY463954.1 - 2, 3 230
          AY463955.1 - 2, 3 230
          AY463956.1 - 2, 3 230
          AY463957.1 - 2, 3 230
          FJ626420.1 - 1, 2 54
          FJ626421.1 - 2, 3 220
          FJ626422.1 - 2, 3 220
          FJ626423.1 - 2, 3 220
          FJ626425.1 - 2, 3 220
          M96944.1 - 2, 3 295

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001109261.
          This siRNA also targets the following transcripts in Mouse: NM_008782, JX065130, M97013.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          humoral immune response
          multicellular organism development
          spermatogenesis
          aging
          organ morphogenesis
          lateral ventricle development
          cerebral cortex development
          adult behavior
          skeletal muscle cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          embryonic cranial skeleton morphogenesis
          negative regulation of histone H3-K9 methylation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0