Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MMP12 SIRNA Assays ›
          Gene Symbol:
          Mmp12
          Gene Name:
          matrix metallopeptidase 12
          Gene Aliases:
          AV378681, Mmel
          Chromosome Location:
          Chr.9: 7344397 - 7360461 on Build GRCm38
          UniGene ID:
          Mm.2055
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 63053

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mmp12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008605.3 NP_032631.3 2 266
          GenBank mRNA AK089295.1 - 235
          AK089309.1 - 2 235
          AK089335.1 - 2 231
          AK089452.1 - 2 235
          AK089509.1 - 3 109
          AK089523.1 - 2 235
          AK090051.1 - 2 237
          AK150596.1 - 2 235
          AK151021.1 - 2 235
          AK151273.1 - 2 235
          AK151933.1 - 2 235
          AK155139.1 - 2 234
          AK155743.1 - 2 235
          AK155747.1 - 2 235
          AK155748.1 - 2 235
          AK155841.1 - 2 293
          AK155865.1 - 2 234
          AK170360.1 - 2 235
          AK170364.1 - 2 235
          AK170500.1 - 3 109
          AK170667.1 - 1 1281
          AK170798.1 - 2 234
          AK170804.1 - 2 234
          AK170976.1 - 2 235
          AK171006.1 - 2 235
          AK171032.1 - 2 234
          AK171036.1 - 2 235
          AK171176.1 - 2 235
          AK171428.1 - 2 235
          AK171435.1 - 1 1282
          AK171447.1 - 2 235
          AK171448.1 - 2 235
          AK171449.1 - 2 235
          AK171450.1 - 2 235
          AK171466.1 - 2 235
          AK171468.1 - 2 235
          AK171470.1 - 2 235
          AK171472.1 - 2 235
          AK171485.1 - 2 235
          AK171518.1 - 2 235
          AK171542.1 - 2 235
          AK171570.1 - 2 235
          BC019135.1 - 2 229 5037674
          CT010377.1 - 2 182
          M82831.1 - 2 182

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053963, BC088120, X98517.

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          wound healing, spreading of epidermal cells
          wound healing
          positive regulation of epithelial cell proliferation involved in wound healing
          metalloendopeptidase activity
          calcium ion binding
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0