Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tarbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009319.3 | NP_033345.2 | 2 | 474 | ||
GenBank mRNA | AK143188.1 | - | 2 | 475 | ||
AV454829.1 | - | 2 | 162 | |||
AV582078.1 | - | 2 | 332 | |||
BC002028.1 | - | 2 | 187 | 3489376 | ||
BI737851.1 | - | 2 | 421 | |||
BQ948575.1 | - | 2 | 215 | |||
U79962.1 | - | 2 | 207 |