Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MIF SIRNA Assays ›
          Gene Symbol:
          Mif
          Gene Name:
          macrophage migration inhibitory factor
          Gene Aliases:
          GIF, Glif
          Chromosome Location:
          Chr.10: 75859353 - 75860250 on Build GRCm38
          UniGene ID:
          Mm.2326
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 68069

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mif
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010798.2 NP_034928.1 3 531
          GenBank mRNA AK008939.1 - 3 525
          AK010691.1 - 3 502
          AK013328.1 - 3 525
          AK152211.1 - 3 512
          BC024895.1 - 3 511 3978685
          BC086928.1 - 3 501 6762214
          BC100605.1 - 3 501 30118109
          L02913.1 - 3 504
          L07607.1 - 3 434
          L10613.1 - 3 533

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          decarboxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          positive regulation of protein phosphorylation
          brain renin-angiotensin system
          immune system process
          positive regulation of acute inflammatory response
          positive regulation of adaptive immune response
          negative regulation of mature B cell apoptotic process
          inflammatory response
          cell surface receptor signaling pathway
          spermatogenesis
          cell aging
          cell proliferation
          negative regulation of gene expression
          positive regulation of protein kinase A signaling
          positive regulation of smooth muscle cell migration
          carboxylic acid metabolic process
          DNA damage response, signal transduction by p53 class mediator
          positive regulation of B cell proliferation
          positive regulation of lipopolysaccharide-mediated signaling pathway
          negative regulation of cellular protein metabolic process
          negative regulation of myeloid cell apoptotic process
          positive regulation of peptidyl-serine phosphorylation
          regulation of cell proliferation
          positive regulation of phosphorylation
          drinking behavior
          negative regulation of apoptotic process
          positive regulation of potassium ion transport
          positive regulation of MAP kinase activity
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          innate immune response
          positive regulation of glycolytic process
          positive regulation of glucose import
          positive regulation of fibroblast proliferation
          positive regulation of axon regeneration
          positive regulation of cytokine secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of immune response
          positive chemotaxis
          positive regulation of prostaglandin secretion involved in immune response
          positive regulation of myeloid leukocyte cytokine production involved in immune response
          protein homotrimerization
          positive regulation of ERK1 and ERK2 cascade
          negative regulation of cell cycle arrest
          positive regulation of arachidonic acid secretion
          negative regulation of cell aging
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of chemokine (C-X-C motif) ligand 2 production
          dopachrome isomerase activity
          receptor binding
          cytokine activity
          cytokine receptor binding
          isomerase activity
          protein complex binding
          chemoattractant activity
          phenylpyruvate tautomerase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0