Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADK SIRNA Assays ›
          Gene Symbol:
          Adk
          Gene Name:
          adenosine kinase
          Gene Aliases:
          2310026J05Rik, 5033405D03Rik, AI255373, AI987814, Ak
          Chromosome Location:
          Chr.14: 21052573 - 21448569 on Build GRCm38
          UniGene ID:
          Mm.188734
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 87737

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 283.00
          Your Price
          Online offer: 274.65
          274.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Adk
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243041.1 NP_001229970.1 7 736
          NM_134079.4 NP_598840.1 7 693
          GenBank mRNA AK032871.1 - 7 651
          AK151863.1 - 7 738
          AK160264.1 - 7 690
          AY540996.1 - 7 635
          AY540997.1 - 6 580
          BC009659.1 - 7 658 3485959

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          purine ribonucleoside salvage
          AMP biosynthetic process
          dATP biosynthetic process
          positive regulation of cardiac muscle hypertrophy
          phosphorylation
          positive regulation of T cell proliferation
          type B pancreatic cell proliferation
          adenosine metabolic process
          nucleotide binding
          adenosine kinase activity
          ATP binding
          kinase activity
          transferase activity
          phosphotransferase activity, alcohol group as acceptor
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline