Search
Search

Galnt7| Gene Symbol: Galnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167981.1 | NP_001161453.1 | 12 | 2405 | ||
| NM_144731.4 | NP_653332.3 | 12 | 2405 | |||
| GenBank mRNA | AK030683.1 | - | 12 | 2353 | ||
| AK033427.1 | - | 12 | 2354 | |||
| AK036523.1 | - | 12 | 2354 | |||
| AK151424.1 | - | 11 | 1996 | |||
| AK151753.1 | - | 11 | 1996 | |||
| AK152205.1 | - | 8 | 1408 | |||
| AK153143.1 | - | 11 | 1996 | |||
| BC007484.1 | - | 8 | 1443 | |||
| BC049907.1 | - | 8 | 1452 | |||
| BC052461.1 | - | 12 | 2381 | 6834581 | ||