Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PICALM SIRNA Assays ›
          Gene Symbol:
          Picalm
          Gene Name:
          phosphatidylinositol binding clathrin assembly protein
          Gene Aliases:
          CALM, CLTH, PAP180, fit-1, fit1, mKIAA4114
          Chromosome Location:
          Chr.7: 90130220 - 90209447 on Build GRCm38
          UniGene ID:
          Mm.235175
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 90943

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Picalm
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252520.1 NP_001239449.1 21 3506
          NM_001252521.1 NP_001239450.1 20 3497
          NM_001252522.1 NP_001239451.1 20 3371
          NM_001252523.1 NP_001239452.1 20 3356
          NM_001252524.1 NP_001239453.1 19 3332
          NM_146194.4 NP_666306.2 21 3521
          GenBank mRNA AK085472.2 - 16 2566
          AK144960.1 - 21 3285
          AK162360.1 - 21 3199
          AK165066.1 - 15 2470
          AK170373.1 - 19 3352
          AK220291.1 - 21 3222
          BC011470.1 - 21 3216 2651109
          BC021491.1 - 19 3281 5345558
          BC023843.1 - 20 3245 5346041
          BC025566.1 - 20 3394 5322606
          BC027116.1 - 17 2793

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001008660, NM_001206946, NM_007166, NM_001206947, BC073961, and others.

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          receptor-mediated endocytosis
          multicellular organism development
          axonogenesis
          cell proliferation
          negative regulation of gene expression
          synaptic vesicle maturation
          vesicle-mediated transport
          endosomal transport
          hemopoiesis
          regulation of endocytosis
          receptor internalization
          regulation of protein localization
          cargo loading into vesicle
          positive regulation of transcription, DNA-templated
          negative regulation of receptor-mediated endocytosis
          clathrin coat assembly
          dendrite morphogenesis
          regulation of protein transport
          iron ion homeostasis
          clathrin-mediated endocytosis
          iron ion import into cell
          positive regulation of neuron death
          positive regulation of beta-amyloid formation
          regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          positive regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
          phospholipid binding
          1-phosphatidylinositol binding
          SH3 domain binding
          clathrin binding
          clathrin heavy chain binding
          clathrin adaptor activity
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0