Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Osbpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148958.2 | NP_683761.1 | 5 | 623 | ||
GenBank mRNA | AK017034.1 | - | 7 | 954 | ||
AK037617.1 | - | 4 | 764 | |||
BC027256.1 | - | 5 | 623 | 2650570 |