Search
Search

Kdm1b| Gene Symbol: Kdm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172262.3 | NP_758466.1 | 2, 3 | 280 | ||
| GenBank mRNA | BC023917.1 | - | 2, 3 | 221 | 5323227 | |
| BY331331.1 | - | 2, 3 | 240 | |||
| CF531857.1 | - | 2, 3 | 228 | |||
| CF733387.1 | - | 1, 2 | 43 | |||