Search
Search

SLC29A2| Gene Symbol: SLC29A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300868.1 | NP_001287797.1 | 10 | 1202 | ||
| NM_001532.2 | NP_001523.2 | 8 | 1001 | |||
| NR_125343.1 | 8 | 1001 | ||||
| XM_011544974.1 | XP_011543276.1 | 10 | 1152 | |||
| XM_017017630.1 | XP_016873119.1 | 8 | 1355 | |||
| XM_017017631.1 | XP_016873120.1 | 8 | 1355 | |||
| XM_017017632.1 | XP_016873121.1 | 10 | 1152 | |||
| XM_017017633.1 | XP_016873122.1 | 8 | 1355 | |||
| XM_017017636.1 | XP_016873125.1 | 8 | 1355 | |||
| XM_017017638.1 | XP_016873127.1 | 6 | 525 | |||
| GenBank mRNA | AF029358.1 | - | 8 | 775 | ||
| AF034102.1 | - | 8 | 1009 | |||
| AF401235.1 | - | 8 | 1009 | |||
| AK057041.1 | - | 9 | 992 | |||
| BC093634.1 | - | 8 | 821 | 7939479 | ||
| BC143335.1 | - | 8 | 821 | |||
| X86681.1 | - | 8 | 767 | |||