Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRP6 SIRNA Assays ›
          Gene Symbol:
          LRP6
          Gene Name:
          LDL receptor related protein 6
          Gene Aliases:
          ADCAD2, STHAG7
          Chromosome Location:
          Chr.12: 12116025 - 12267021 on Build GRCh38
          UniGene ID:
          Hs.584775
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS106153
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRP6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002336.2 NP_002327.2 12 2708
          XM_006719078.3 XP_006719141.1 12 2852
          XM_011520671.2 XP_011518973.1 11 2458
          XM_017019304.1 XP_016874793.1 10 2526
          GenBank mRNA AB209683.1 - 11 2160
          AF074264.1 - 12 2643
          AK074902.1 - 2 245
          BC117136.1 - 12 2600 40125687
          BC126405.1 - 12 2600 8992121
          BC143725.1 - 12 2600 9052239

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          negative regulation of protein phosphorylation
          pericardium morphogenesis
          negative regulation of protein kinase activity
          positive regulation of cytosolic calcium ion concentration
          chemical synaptic transmission
          embryonic pattern specification
          neural crest formation
          neural crest cell differentiation
          Wnt signaling pathway
          single organismal cell-cell adhesion
          cerebellum morphogenesis
          thalamus development
          cerebral cortex development
          midbrain-hindbrain boundary development
          negative regulation of smooth muscle cell apoptotic process
          external genitalia morphogenesis
          odontogenesis of dentin-containing tooth
          response to peptide hormone
          Wnt signaling pathway involved in dorsal/ventral axis specification
          canonical Wnt signaling pathway involved in neural crest cell differentiation
          canonical Wnt signaling pathway involved in regulation of cell proliferation
          positive regulation of cell cycle
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          palate development
          convergent extension
          embryonic retina morphogenesis in camera-type eye
          canonical Wnt signaling pathway
          face morphogenesis
          trachea cartilage morphogenesis
          cellular response to cholesterol
          dopaminergic neuron differentiation
          negative regulation of protein serine/threonine kinase activity
          protein localization to plasma membrane
          primitive streak formation
          negative regulation of canonical Wnt signaling pathway
          receptor-mediated endocytosis of low-density lipoprotein particle involved in cholesterol transport
          Wnt signaling pathway involved in somitogenesis
          axis elongation involved in somitogenesis
          positive regulation of canonical Wnt signaling pathway
          toxin transport
          beta-catenin destruction complex disassembly
          midbrain dopaminergic neuron differentiation
          Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
          positive regulation of Wnt signaling pathway involved in dorsal/ventral axis specification
          low-density lipoprotein receptor activity
          receptor binding
          frizzled binding
          protein binding
          Wnt-protein binding
          kinase inhibitor activity
          toxin transporter activity
          apolipoprotein binding
          identical protein binding
          protein homodimerization activity
          Wnt-activated receptor activity
          coreceptor activity involved in Wnt signaling pathway
          coreceptor activity involved in canonical Wnt signaling pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0