Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other C2CD5 SIRNA Assays ›
          Gene Symbol:
          C2CD5
          Gene Name:
          C2 calcium dependent domain containing 5
          Gene Aliases:
          CDP138, KIAA0528
          Chromosome Location:
          Chr.12: 22448546 - 22544546 on Build GRCh38
          UniGene ID:
          Hs.271014
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS114869
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: C2CD5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286173.1 NP_001273102.1 26, 27 3127
          NM_001286174.1 NP_001273103.1 24, 25 3261
          NM_001286175.1 NP_001273104.1 26, 27 3130
          NM_001286176.1 NP_001273105.1 25, 26 3121
          NM_001286177.1 NP_001273106.1 25, 26 3094
          NM_014802.2 NP_055617.1 23, 24 2968
          XM_005253538.2 XP_005253595.1 23, 24 2903
          XM_005253539.3 XP_005253596.1 17, 18 1813
          XM_006719170.2 XP_006719233.1 24, 25 2981
          XM_017020264.1 XP_016875753.1 29, 30 3413
          XM_017020265.1 XP_016875754.1 29, 30 3383
          XM_017020266.1 XP_016875755.1 28, 29 3362
          XM_017020267.1 XP_016875756.1 27, 28 3341
          XM_017020268.1 XP_016875757.1 26, 27 3275
          XM_017020269.1 XP_016875758.1 27, 28 3260
          XM_017020270.1 XP_016875759.1 28, 29 3221
          XM_017020271.1 XP_016875760.1 28, 29 3194
          XM_017020272.1 XP_016875761.1 26, 27 3122
          XM_017020273.1 XP_016875762.1 25, 26 3071
          XM_017020274.1 XP_016875763.1 25, 26 3053
          XM_017020275.1 XP_016875764.1 26, 27 3041
          XM_017020276.1 XP_016875765.1 25, 26 3026
          XM_017020277.1 XP_016875766.1 24, 25 3005
          XM_017020278.1 XP_016875767.1 24, 25 2969
          XM_017020279.1 XP_016875768.1 21, 22 2136
          GenBank mRNA AB011100.3 - 22, 23 2721
          AK295862.1 - 26, 27 3075
          AK299353.1 - 24, 25 2968
          AY166851.1 - 23, 24 2940
          BC042498.1 - 22, 23 2727 5174801
          BC053885.1 - 18, 19 2192
          BC117143.1 - 25, 26 2957 40125694
          BC143859.1 - 25, 26 2984 9052375
          BC143860.1 - 25, 26 2957 9052376
          BC143878.1 - 26, 27 2990 9052394
          BP418369.1 - 1, 2 81
          BX648560.1 - 6, 7 585

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          vesicle fusion
          positive regulation of glucose transport
          positive regulation of vesicle fusion
          cellular response to insulin stimulus
          insulin receptor signaling pathway via phosphatidylinositol 3-kinase
          intracellular protein transmembrane transport
          protein localization to plasma membrane
          positive regulation of protein targeting to membrane
          positive regulation of glucose import in response to insulin stimulus
          calcium ion binding
          calcium-dependent phospholipid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline