This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
RAMP3
- Gene Name:
-
receptor (G protein-coupled) activity modifying protein 3
- Gene Aliases:
-
-
- Chromosome Location:
-
Chr.7: 45157768 - 45184250 on Build GRCh38
- UniGene ID:
-
Hs.25691
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
RAMP3 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_005856.2 |
NP_005847.1 |
2, 3
|
252
|
|
| XM_006715631.3 |
XP_006715694.1 |
4, 5
|
575
|
|
| XM_017011666.1 |
XP_016867155.1 |
2, 3
|
616
|
|
| GenBank mRNA |
AJ001016.1 |
- |
2, 3
|
220
|
|
| AK222469.1 |
- |
2, 3
|
216
|
|
| AK311871.1 |
- |
2, 3
|
191
|
|
| AY265459.1 |
- |
2, 3
|
191
|
|
| BC022304.1 |
- |
2, 3
|
219
|
4717934 |
| BC053852.1 |
- |
2, 3
|
218
|
6197363 |
| BP347072.1 |
- |
2, 3
|
252
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System