Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNIP1 SIRNA Assays ›
          Gene Symbol:
          TNIP1
          Gene Name:
          TNFAIP3 interacting protein 1
          Gene Aliases:
          ABIN-1, NAF1, VAN, nip40-1
          Chromosome Location:
          Chr.5: 151029943 - 151087660 on Build GRCh38
          UniGene ID:
          Hs.355141
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS115778
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNIP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252385.1 NP_001239314.1 15 1779
          NM_001252386.1 NP_001239315.1 14 1607
          NM_001252390.1 NP_001239319.1 15 1644
          NM_001252391.1 NP_001239320.1 15 1676
          NM_001252392.1 NP_001239321.1 15 1676
          NM_001252393.1 NP_001239322.1 15 1779
          NM_001258454.1 NP_001245383.1 15 1779
          NM_001258455.1 NP_001245384.1 14 1574
          NM_001258456.1 NP_001245385.1 14 1574
          NM_006058.4 NP_006049.3 15 1779
          XM_005268355.1 XP_005268412.1 15 1692
          XM_006714751.1 XP_006714814.1 15 1692
          XM_006714752.2 XP_006714815.1 15 1692
          XM_017008945.1 XP_016864434.1 15 1692
          XM_017008946.1 XP_016864435.1 15 1658
          XM_017008947.1 XP_016864436.1 16 2161
          XM_017008948.1 XP_016864437.1 14 1953
          XM_017008949.1 XP_016864438.1 15 1692
          XM_017008950.1 XP_016864439.1 15 1692
          XM_017008951.1 XP_016864440.1 14 1953
          GenBank mRNA AB177543.1 - 14 1538
          AB177544.1 - 14 1538
          AB252970.1 - 15 1648
          AB252971.1 - 15 1648
          AB252972.1 - 15 1769
          AB252973.1 - 15 1769
          AB252976.1 - 13 1402
          AB252977.1 - 13 1402
          AB252978.1 - 13 1402
          AB252979.1 - 13 1402
          AB252980.1 - 13 1402
          AB252981.1 - 13 1402
          AB252982.1 - 15 1648
          AJ011895.1 - 15 1648
          AJ011896.1 - 15 1648
          AK074400.1 - 15 1710
          AK290999.1 - 15 1645
          AK298685.1 - 14 1601
          AK299975.1 - 14 1539
          AK303573.1 - 15 1729
          AY012155.1 - 15 1795
          BC012133.1 - 15 1685 4549542
          BC014008.1 - 15 1626 4040118
          BX640647.1 - 14 1528
          D30755.2 - 14 1538
          U39403.1 - 11 1134

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MyD88-dependent toll-like receptor signaling pathway
          translation
          proteolysis
          defense response
          inflammatory response
          leukocyte cell-cell adhesion
          glycoprotein biosynthetic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of viral genome replication
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of inflammatory response
          negative regulation of ERK1 and ERK2 cascade
          modulation by symbiont of host I-kappaB kinase/NF-kappaB cascade
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          mitogen-activated protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0