Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMCC1 SIRNA Assays ›
          Gene Symbol:
          TMCC1
          Gene Name:
          transmembrane and coiled-coil domain family 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 129647792 - 129893605 on Build GRCh38
          UniGene ID:
          Hs.477547
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS118050
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001017395.3 NP_001017395.2 4 1860
          NM_001128224.2 NP_001121696.1 4 1261
          NR_033361.1 2 1406
          XM_006713542.3 XP_006713605.1 5 1973
          XM_006713543.3 XP_006713606.1 5 1970
          XM_006713544.1 XP_006713607.1 5 2098
          XM_006713550.3 XP_006713613.1 5 1531
          XM_006713552.3 XP_006713615.1 2 1153
          XM_006713553.3 XP_006713616.1 2 2445
          XM_011512572.2 XP_011510874.1 6 2133
          XM_011512573.2 XP_011510875.1 5 2712
          XM_011512576.2 XP_011510878.1 5 1966
          XM_011512579.2 XP_011510881.1 4 1721
          XM_011512580.2 XP_011510882.1 7 1858
          XM_011512581.2 XP_011510883.1 6 1695
          XM_011512582.1 XP_011510884.1 2 1487
          XM_011512583.2 XP_011510885.1 4 1264
          XM_011512584.2 XP_011510886.1 6 1583
          XM_017005931.1 XP_016861420.1 5 1881
          XM_017005932.1 XP_016861421.1 6 2120
          XM_017005933.1 XP_016861422.1 4 1917
          XM_017005934.1 XP_016861423.1 8 1931
          XM_017005935.1 XP_016861424.1 9 2020
          XM_017005936.1 XP_016861425.1 6 1694
          XM_017005937.1 XP_016861426.1 6 1628
          XM_017005938.1 XP_016861427.1 8 1994
          XM_017005939.1 XP_016861428.1 7 1888
          XM_017005940.1 XP_016861429.1 4 1266
          XM_017005941.1 XP_016861430.1 4 1392
          GenBank mRNA AB018322.1 - 1 427
          AK124671.1 - 2 1407
          AK291448.1 - 4 1679
          AK293408.1 - 2 943
          BC030619.2 - 2 1324
          BC039859.1 - 4 1767 5527623
          BC047517.1 - 2 1324
          BX640712.1 - 2 914
          CR749206.1 - 2 1386

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0