Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMUG1 SIRNA Assays ›
          Gene Symbol:
          SMUG1
          Gene Name:
          single-strand-selective monofunctional uracil-DNA glycosylase 1
          Gene Aliases:
          FDG, HMUDG, UNG3
          Chromosome Location:
          Chr.12: 54171815 - 54188994 on Build GRCh38
          UniGene ID:
          Hs.632721
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS119001
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMUG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243787.1 NP_001230716.1 4 918
          NM_001243788.1 NP_001230717.1 3 834
          NM_014311.2 NP_055126.1 4 921
          XM_006719319.3 XP_006719382.1 5 1020
          XM_006719320.3 XP_006719383.1 4 1127
          XM_006719321.3 XP_006719384.1 3 872
          XM_011538109.2 XP_011536411.1 5 1191
          XM_011538110.2 XP_011536412.1 5 1194
          XM_011538111.2 XP_011536413.1 4 1095
          XM_011538112.2 XP_011536414.1 4 1097
          XM_011538113.2 XP_011536415.1 4 960
          XM_011538114.2 XP_011536416.1 4 1116
          XM_011538115.2 XP_011536417.1 4 990
          XM_011538116.2 XP_011536418.1 4 930
          XM_011538117.2 XP_011536419.1 4 906
          XM_011538118.2 XP_011536420.1 4 951
          XM_011538119.2 XP_011536421.1 4 893
          XM_011538120.2 XP_011536422.1 3 871
          XM_011538121.2 XP_011536423.1 5 1195
          XM_017019113.1 XP_016874602.1 5 1185
          XM_017019114.1 XP_016874603.1 3 1268
          XM_017019115.1 XP_016874604.1 4 993
          XM_017019116.1 XP_016874605.1 6 1447
          GenBank mRNA AF125182.1 - 2 803
          AK001235.1 - 4 900
          AK001789.1 - 3 1925
          AK091468.1 - 4 628
          AK222473.1 - 4 897
          AK290274.1 - 3 826
          KU178579.1 - 2 771

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          base-excision repair
          depyrimidination
          oxidized pyrimidine nucleobase lesion DNA N-glycosylase activity
          DNA binding
          uracil DNA N-glycosylase activity
          protein binding
          single-strand selective uracil DNA N-glycosylase activity
          DNA N-glycosylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0