Search
Search

ST8SIA3| Gene Symbol: ST8SIA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015879.2 | NP_056963.2 | 2 | 574 | ||
| GenBank mRNA | AB208996.1 | - | 2 | 578 | ||
| AF003092.2 | - | 2 | 289 | |||
| AF004668.2 | - | 2 | 2240 | |||
| AK289517.1 | - | 2 | 352 | |||
| AK313047.1 | - | 2 | 300 | |||
| BC074909.2 | - | 2 | 265 | 30915256 | ||