Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171740.2 | NP_001165211.1 | 2 | 676 | ||
NM_001171741.2 | NP_001165212.1 | 2 | 471 | |||
NM_016210.4 | NP_057294.2 | 3 | 765 | |||
XM_011533782.2 | XP_011532084.1 | 3 | 741 | |||
XM_011533783.2 | XP_011532085.1 | 2 | 631 | |||
XM_011533784.2 | XP_011532086.1 | 3 | 519 | |||
XM_011533785.2 | XP_011532087.1 | 2 | 457 | |||
XM_017006545.1 | XP_016862034.1 | 3 | 741 | |||
GenBank mRNA | AF188706.1 | - | 2 | 436 | ||
AK124438.1 | - | 2 | 516 | |||
AK313128.1 | - | 3 | 639 | |||
AL833938.1 | - | 3 | 724 | |||
BC034766.1 | - | 2 | 549 | 5177784 |