Search
Search

KIF9| Gene Symbol: KIF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134878.1 | NP_001128350.1 | 11 | 1635 | ||
| NM_022342.4 | NP_071737.1 | 10 | 1389 | |||
| NM_182902.3 | NP_878905.2 | 10 | 1389 | |||
| XM_005265388.4 | XP_005265445.1 | 10 | 1164 | |||
| XM_005265389.4 | XP_005265446.1 | 10 | 1164 | |||
| XM_006713291.3 | XP_006713354.1 | 9 | 939 | |||
| XM_011534003.2 | XP_011532305.1 | 11 | 1364 | |||
| XM_011534004.2 | XP_011532306.1 | 11 | 1588 | |||
| XM_011534005.2 | XP_011532307.1 | 9 | 896 | |||
| XM_011534006.2 | XP_011532308.1 | 9 | 1282 | |||
| XM_011534007.2 | XP_011532309.1 | 3 | 163 | |||
| XM_017007029.1 | XP_016862518.1 | 11 | 1360 | |||
| XM_017007030.1 | XP_016862519.1 | 10 | 1363 | |||
| XM_017007031.1 | XP_016862520.1 | 9 | 896 | |||
| GenBank mRNA | AF311212.1 | - | 10 | 1092 | ||
| AK023894.1 | - | 10 | 1092 | |||
| AK292547.1 | - | 10 | 1083 | |||
| AK302070.1 | - | 9 | 1033 | |||
| AK302974.1 | - | 11 | 1146 | |||
| AK314418.1 | - | 11 | 1366 | |||
| BC030657.2 | - | 10 | 1140 | 4829657 | ||