Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLG2 SIRNA Assays ›
          Gene Symbol:
          DLG2
          Gene Name:
          discs large MAGUK scaffold protein 2
          Gene Aliases:
          PPP1R58, PSD-93, PSD93, chapsyn-110
          Chromosome Location:
          Chr.11: 83455009 - 85628534 on Build GRCh38
          UniGene ID:
          Hs.367656
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS138273
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DLG2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142699.1 NP_001136171.1 28 5877
          NM_001142700.1 NP_001136172.1 20 4912
          NM_001142702.1 NP_001136174.1 11 3954
          NM_001206769.1 NP_001193698.1 22 5561
          NM_001300983.1 NP_001287912.1 23 5499
          NM_001364.3 NP_001355.2 23 5553
          XM_005273810.4 XP_005273867.1 22 5196
          XM_005273811.4 XP_005273868.1 22 5142
          XM_011544778.2 XP_011543080.1 29 7168
          XM_011544780.2 XP_011543082.1 29 7114
          XM_011544782.2 XP_011543084.1 24 5639
          XM_011544788.2 XP_011543090.1 21 4986
          XM_017017254.1 XP_016872743.1 30 7210
          XM_017017255.1 XP_016872744.1 29 7171
          XM_017017256.1 XP_016872745.1 29 6700
          XM_017017257.1 XP_016872746.1 28 7632
          XM_017017258.1 XP_016872747.1 27 6808
          XM_017017259.1 XP_016872748.1 27 6529
          XM_017017260.1 XP_016872749.1 26 7461
          XM_017017261.1 XP_016872750.1 28 6958
          XM_017017262.1 XP_016872751.1 23 5663
          XM_017017263.1 XP_016872752.1 25 5681
          XM_017017264.1 XP_016872753.1 24 5858
          XM_017017265.1 XP_016872754.1 24 5855
          XM_017017266.1 XP_016872755.1 22 5565
          XM_017017267.1 XP_016872756.1 24 5763
          XM_017017268.1 XP_016872757.1 24 5584
          XM_017017269.1 XP_016872758.1 24 6285
          XM_017017270.1 XP_016872759.1 24 5585
          XM_017017271.1 XP_016872760.1 23 5762
          XM_017017272.1 XP_016872761.1 23 5759
          XM_017017273.1 XP_016872762.1 22 5514
          XM_017017274.1 XP_016872763.1 24 5709
          XM_017017275.1 XP_016872764.1 21 5464
          XM_017017276.1 XP_016872765.1 23 6187
          XM_017017277.1 XP_016872766.1 23 5507
          XM_017017278.1 XP_016872767.1 21 5411
          XM_017017279.1 XP_016872768.1 23 5429
          XM_017017280.1 XP_016872769.1 23 6106
          XM_017017281.1 XP_016872770.1 22 5606
          XM_017017283.1 XP_016872772.1 22 5411
          XM_017017284.1 XP_016872773.1 22 5083
          XM_017017285.1 XP_016872774.1 22 6033
          XM_017017286.1 XP_016872775.1 20 4947
          XM_017017287.1 XP_016872776.1 13 3967
          XM_017017288.1 XP_016872777.1 12 3806
          XM_017017289.1 XP_016872778.1 12 5381
          XM_017017290.1 XP_016872779.1 12 4026
          XM_017017291.1 XP_016872780.1 11 3773
          XM_017017292.1 XP_016872781.1 11 5285
          GenBank mRNA AK125151.1 - 1 983
          AK125152.1 - 1 506
          BC104969.1 - 1 74
          BC110844.1 - 1 329
          BX648386.1 - 1 1380
          CR933674.1 - 11 3923

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_011807, NM_001243047, NM_001243046, AK084856, AK032411.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          nervous system development
          negative regulation of phosphatase activity
          sensory perception of pain
          receptor clustering
          establishment or maintenance of epithelial cell apical/basal polarity
          GMP metabolic process
          GDP metabolic process
          receptor localization to synapse
          guanylate kinase activity
          protein binding
          kinase binding
          ionotropic glutamate receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0