Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GPHN SIRNA Assays ›
Gene Symbol:
GPHN
Gene Name:
gephyrin
Gene Aliases:
GEPH, GPH, GPHRYN, HKPX1, MOCODC
Chromosome Location:
Chr.14: 66507407 - 67735831 on Build GRCh38
UniGene ID:
Hs.208765
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS173497
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GPHN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024218.1
NP_001019389.1
16
2689
NM_020806.4
NP_065857.1
17
2788
XM_005267254.3
XP_005267311.1
17
2041
XM_011536340.2
XP_011534642.1
20
2429
XM_011536342.2
XP_011534644.1
19
2390
XM_011536343.2
XP_011534645.1
19
2357
XM_011536344.2
XP_011534646.1
19
2330
XM_011536345.2
XP_011534647.1
18
2300
XM_011536346.2
XP_011534648.1
18
2092
XM_011536347.2
XP_011534649.1
17
1687
XM_017020913.1
XP_016876402.1
20
2420
XM_017020914.1
XP_016876403.1
19
2372
XM_017020915.1
XP_016876404.1
19
2321
XM_017020916.1
XP_016876405.1
18
2282
XM_017020917.1
XP_016876406.1
18
2273
XM_017020918.1
XP_016876407.1
18
2258
XM_017020919.1
XP_016876408.1
17
2234
XM_017020920.1
XP_016876409.1
18
2223
XM_017020921.1
XP_016876410.1
20
2492
XM_017020922.1
XP_016876411.1
21
1958
XM_017020923.1
XP_016876412.1
18
2363
XM_017020924.1
XP_016876413.1
20
2423
XM_017020926.1
XP_016876415.1
19
2351
GenBank mRNA
AB037806.1
-
16
2689
AF272663.1
-
16
2689
AJ272033.1
-
16
1568
AJ272343.1
-
17
1707
AK294532.1
-
19
1730
AK295577.1
-
15
1565
AK297447.1
-
15
1490
AK315569.1
-
17
2037
BC030016.2
-
17
1679
4826821
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
Mo-molybdopterin cofactor biosynthetic process
molybdenum incorporation into molybdenum-molybdopterin complex
molybdopterin cofactor biosynthetic process
glycine receptor clustering
gamma-aminobutyric acid receptor clustering
protein binding
ATP binding
metal ion binding
molybdopterin adenylyltransferase activity
molybdopterin molybdotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-k2gqz:80/100.66.79.23:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0