Search
Search

B3GNT3| Gene Symbol: B3GNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014256.3 | NP_055071.2 | 3 | 1037 | ||
| XM_011527626.2 | XP_011525928.1 | 3 | 996 | |||
| GenBank mRNA | AB015630.1 | - | 3 | 1007 | ||
| AB049585.1 | - | 3 | 992 | |||
| AF293973.1 | - | 2 | 903 | |||
| AJ130847.1 | - | 2 | 890 | |||
| AJ278961.1 | - | 2 | 896 | |||
| AK075434.1 | - | 4 | 587 | |||
| AK075466.1 | - | 3 | 1012 | |||
| AK225421.1 | - | 3 | 1004 | |||
| AK225478.1 | - | 3 | 983 | |||
| AK314323.1 | - | 3 | 1008 | |||
| AY358955.1 | - | 3 | 992 | |||
| BC066876.1 | - | 3 | 1009 | 6971715 | ||
| BC067423.1 | - | 3 | 988 | 6971716 | ||