Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085357.1 | NP_001078826.1 | 4 | 780 | ||
NM_181780.3 | NP_861445.3 | 5 | 924 | |||
XM_011512447.2 | XP_011510749.1 | 6 | 1036 | |||
XM_017005748.1 | XP_016861237.1 | 6 | 999 | |||
GenBank mRNA | AJ717664.1 | - | 5 | 721 | ||
AK131204.1 | - | 5 | 713 | |||
AK292692.1 | - | 5 | 903 | |||
AW294080.1 | - | 1 | 344 | |||
AY599411.1 | - | 4 | 577 | |||
AY606805.1 | - | 5 | 721 | |||
BC107091.1 | - | 4 | 577 | 40015588 | ||
BC107092.1 | - | 5 | 719 | |||
DQ198368.1 | - | 4 | 577 |