Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLG2 SIRNA Assays ›
Gene Symbol:
DLG2
Gene Name:
discs large MAGUK scaffold protein 2
Gene Aliases:
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location:
Chr.11: 83455009 - 85628534 on Build GRCh38
UniGene ID:
Hs.367656
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS176190
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142699.1
NP_001136171.1
13
1435
NM_001142700.1
NP_001136172.1
6
680
NM_001206769.1
NP_001193698.1
7
1119
NM_001300983.1
NP_001287912.1
8
1111
NM_001364.3
NP_001355.2
8
1111
XM_005273810.4
XP_005273867.1
7
754
XM_005273811.4
XP_005273868.1
7
754
XM_011544778.2
XP_011543080.1
14
2726
XM_011544780.2
XP_011543082.1
14
2726
XM_011544782.2
XP_011543084.1
9
1197
XM_011544788.2
XP_011543090.1
7
754
XM_017017254.1
XP_016872743.1
14
2726
XM_017017255.1
XP_016872744.1
13
2687
XM_017017256.1
XP_016872745.1
13
2216
XM_017017257.1
XP_016872746.1
12
3148
XM_017017258.1
XP_016872747.1
11
2324
XM_017017259.1
XP_016872748.1
12
2141
XM_017017260.1
XP_016872749.1
11
3073
XM_017017261.1
XP_016872750.1
14
2726
XM_017017262.1
XP_016872751.1
7
1179
XM_017017263.1
XP_016872752.1
9
1197
XM_017017264.1
XP_016872753.1
8
1374
XM_017017265.1
XP_016872754.1
8
1371
XM_017017266.1
XP_016872755.1
7
1177
XM_017017267.1
XP_016872756.1
8
1279
XM_017017268.1
XP_016872757.1
8
1100
XM_017017269.1
XP_016872758.1
8
1801
XM_017017270.1
XP_016872759.1
9
1197
XM_017017271.1
XP_016872760.1
8
1374
XM_017017272.1
XP_016872761.1
8
1371
XM_017017273.1
XP_016872762.1
7
1186
XM_017017274.1
XP_016872763.1
8
1279
XM_017017275.1
XP_016872764.1
7
1178
XM_017017276.1
XP_016872765.1
8
1799
XM_017017277.1
XP_016872766.1
7
1023
XM_017017278.1
XP_016872767.1
7
1179
XM_017017279.1
XP_016872768.1
9
1197
XM_017017280.1
XP_016872769.1
9
1874
XM_017017281.1
XP_016872770.1
8
1374
XM_017017282.1
XP_016872771.1
7
754
XM_017017283.1
XP_016872772.1
7
1023
XM_017017284.1
XP_016872773.1
6
599
XM_017017285.1
XP_016872774.1
8
1801
XM_017017286.1
XP_016872775.1
4
463
GenBank mRNA
AB209252.1
-
8
1086
AK126776.1
-
6
680
AK127150.1
-
7
1117
AK294375.1
-
7
642
AK295094.1
-
8
1111
AK295624.1
-
7
734
AK299797.1
-
8
869
CR749820.1
-
13
1435
KJ535093.1
-
13
1318
U32376.1
-
8
1084
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
nervous system development
negative regulation of phosphatase activity
sensory perception of pain
receptor clustering
establishment or maintenance of epithelial cell apical/basal polarity
GMP metabolic process
GDP metabolic process
receptor localization to synapse
guanylate kinase activity
protein binding
kinase binding
ionotropic glutamate receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0