Search
Search

DPYSL3| Gene Symbol: DPYSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197294.1 | NP_001184223.1 | 5 | 1038 | ||
| NM_001387.2 | NP_001378.1 | 5 | 608 | |||
| XM_011537574.1 | XP_011535876.1 | 6 | 992 | |||
| GenBank mRNA | BC039006.1 | - | 5 | 574 | 5725550 | |
| BC077077.1 | - | 5 | 1038 | 6177053 | ||
| D78014.1 | - | 5 | 608 | |||
| EU007906.1 | - | 5 | 840 | |||
| Y07818.1 | - | 5 | 629 | |||