Search
Search

TSPAN12| Gene Symbol: TSPAN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012338.3 | NP_036470.1 | 8 | 1252 | ||
| XM_005250239.2 | XP_005250296.1 | 9 | 1253 | |||
| XM_011515993.1 | XP_011514295.1 | 8 | 1069 | |||
| XM_011515994.1 | XP_011514296.1 | 8 | 987 | |||
| XM_017011912.1 | XP_016867401.1 | 9 | 1158 | |||
| XM_017011913.1 | XP_016867402.1 | 7 | 1200 | |||
| XM_017011914.1 | XP_016867403.1 | 4 | 759 | |||
| GenBank mRNA | AF124522.1 | - | 7 | 943 | ||
| AK299247.1 | - | 5 | 747 | |||
| AK307161.1 | - | 4 | 674 | |||
| AK312239.1 | - | 8 | 1091 | |||
| AY358703.1 | - | 8 | 1085 | |||
| BC031265.1 | - | 8 | 1250 | 5275953 | ||
| BF338835.1 | - | 2 | 328 | |||