Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019091.3 | NP_061964.3 | 6, 7 | 1061 | ||
XM_011511689.2 | XP_011509991.1 | 7, 8 | 1170 | |||
GenBank mRNA | AF286162.1 | - | 6, 7 | 900 | ||
AK000074.1 | - | 4, 5 | 464 | |||
AK074209.1 | - | 6, 7 | 1036 | |||
AK314459.1 | - | 6, 7 | 1065 | |||
BC044567.1 | - | 6, 7 | 755 | 5262097 | ||
DQ786259.1 | - | 3, 4 | 378 | |||
DQ786318.1 | - | 2, 3 | 218 |