Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS186235
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
7
2330
NM_001174093.1
NP_001167564.1
6
1990
NM_001174094.1
NP_001167565.1
7
2327
NM_001174095.1
NP_001167566.1
6
1849
NM_001174096.1
NP_001167567.1
7
2053
NM_001323638.1
NP_001310567.1
11
2700
NM_001323641.1
NP_001310570.1
7
2280
NM_001323642.1
NP_001310571.1
9
2582
NM_001323643.1
NP_001310572.1
9
2360
NM_001323644.1
NP_001310573.1
6
2079
NM_001323645.1
NP_001310574.1
8
2423
NM_001323646.1
NP_001310575.1
10
2661
NM_001323647.1
NP_001310576.1
8
2193
NM_001323648.1
NP_001310577.1
7
2133
NM_001323649.1
NP_001310578.1
8
2196
NM_001323650.1
NP_001310579.1
10
2428
NM_001323651.1
NP_001310580.1
8
2096
NM_001323652.1
NP_001310581.1
9
2245
NM_001323653.1
NP_001310582.1
9
2352
NM_001323654.1
NP_001310583.1
9
2239
NM_001323655.1
NP_001310584.1
9
2349
NM_001323656.1
NP_001310585.1
7
2024
NM_001323657.1
NP_001310586.1
7
2027
NM_001323658.1
NP_001310587.1
7
1866
NM_001323659.1
NP_001310588.1
7
2084
NM_001323660.1
NP_001310589.1
7
2081
NM_001323661.1
NP_001310590.1
8
2227
NM_001323662.1
NP_001310591.1
9
2629
NM_001323663.1
NP_001310592.1
10
2568
NM_001323664.1
NP_001310593.1
9
2552
NM_001323665.1
NP_001310594.1
8
2470
NM_001323666.1
NP_001310595.1
7
2410
NM_001323671.1
NP_001310600.1
8
2306
NM_001323672.1
NP_001310601.1
9
2358
NM_001323673.1
NP_001310602.1
8
2303
NM_001323674.1
NP_001310603.1
5
1828
NM_001323675.1
NP_001310604.1
5
1938
NM_001323676.1
NP_001310605.1
7
2163
NM_001323677.1
NP_001310606.1
7
2160
NM_001323678.1
NP_001310607.1
5
2105
NM_030751.5
NP_110378.3
7
2050
XM_006717498.2
XP_006717561.1
10
2488
XM_011519643.2
XP_011517945.1
10
2791
XM_017016597.1
XP_016872086.1
10
2390
XM_017016598.1
XP_016872087.1
10
5520
XM_017016599.1
XP_016872088.1
9
5358
XM_017016600.1
XP_016872089.1
11
5599
XM_017016601.1
XP_016872090.1
10
5517
XM_017016602.1
XP_016872091.1
11
5596
XM_017016603.1
XP_016872092.1
10
5434
GenBank mRNA
AK091478.1
-
6
1950
AK294775.1
-
7
2245
AK296244.1
-
7
1973
AK299541.1
-
4
1587
AK299666.1
-
7
2084
AK300830.1
-
6
1797
AK307510.1
-
8
2056
AK314683.1
-
7
2011
AL831979.1
-
9
2308
BC009572.1
-
7
2006
BC112392.1
-
7
2006
40036600
BX647794.1
-
11
2701
M81699.1
-
1
922
U12170.1
-
7
2011
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0