Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EIF4G3 SIRNA Assays ›
Gene Symbol:
EIF4G3
Gene Name:
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases:
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location:
Chr.1: 20806292 - 21177320 on Build GRCh38
UniGene ID:
Hs.467084
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS189494
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EIF4G3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198801.1
NP_001185730.1
18
3119
NM_001198802.1
NP_001185731.1
18
3029
NM_003760.4
NP_003751.2
15
2645
XM_011542356.2
XP_011540658.1
21
3719
XM_011542361.2
XP_011540663.1
20
3608
XM_011542370.1
XP_011540672.1
21
3722
XM_011542371.1
XP_011540673.1
19
3464
XM_011542374.1
XP_011540676.1
18
3431
XM_011542375.2
XP_011540677.1
18
5019
XM_011542381.2
XP_011540683.1
8
1105
XM_017002672.1
XP_016858161.1
22
5257
XM_017002673.1
XP_016858162.1
20
3701
XM_017002674.1
XP_016858163.1
20
3698
XM_017002675.1
XP_016858164.1
20
3689
XM_017002676.1
XP_016858165.1
19
3668
XM_017002677.1
XP_016858166.1
19
3545
XM_017002678.1
XP_016858167.1
20
3611
XM_017002679.1
XP_016858168.1
20
3608
XM_017002680.1
XP_016858169.1
17
2638
XM_017002681.1
XP_016858170.1
20
3605
XM_017002682.1
XP_016858171.1
19
3587
XM_017002683.1
XP_016858172.1
19
3587
XM_017002684.1
XP_016858173.1
20
5113
XM_017002685.1
XP_016858174.1
19
3578
XM_017002686.1
XP_016858175.1
19
3575
XM_017002687.1
XP_016858176.1
19
3575
XM_017002688.1
XP_016858177.1
16
2604
XM_017002689.1
XP_016858178.1
21
3722
XM_017002690.1
XP_016858179.1
19
3572
XM_017002691.1
XP_016858180.1
18
3557
XM_017002692.1
XP_016858181.1
18
3554
XM_017002693.1
XP_016858182.1
17
3425
XM_017002694.1
XP_016858183.1
19
3497
XM_017002695.1
XP_016858184.1
19
3494
XM_017002696.1
XP_016858185.1
18
3476
XM_017002697.1
XP_016858186.1
18
3473
XM_017002698.1
XP_016858187.1
18
3464
XM_017002699.1
XP_016858188.1
20
3611
XM_017002700.1
XP_016858189.1
17
3443
XM_017002701.1
XP_016858190.1
17
3440
XM_017002702.1
XP_016858191.1
17
3374
XM_017002703.1
XP_016858192.1
15
2535
XM_017002704.1
XP_016858193.1
19
3587
XM_017002705.1
XP_016858194.1
20
2882
XM_017002706.1
XP_016858195.1
19
2771
XM_017002707.1
XP_016858196.1
17
2714
XM_017002708.1
XP_016858197.1
16
2603
XM_017002709.1
XP_016858198.1
18
3466
XM_017002710.1
XP_016858199.1
21
3722
XM_017002711.1
XP_016858200.1
20
3611
GenBank mRNA
AB209119.1
-
18
2988
AF012072.2
-
15
2645
AK294883.1
-
9
1892
AK302873.1
-
6
886
BC094683.1
-
13
1652
6188273
BC136643.1
-
15
2568
9020633
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
translation initiation factor
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0