Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD1E SIRNA Assays ›
Gene Symbol:
CD1E
Gene Name:
CD1e molecule
Gene Aliases:
CD1A, R2
Chromosome Location:
Chr.1: 158353157 - 158357554 on Build GRCh38
UniGene ID:
Hs.249217
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS189928
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CD1E
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042583.2
NP_001036048.1
5
1217
NM_001042584.2
NP_001036049.1
5
1113
NM_001042585.2
NP_001036050.1
5
1052
NM_001042586.2
NP_001036051.1
3
650
NM_001042587.2
NP_001036052.1
3
546
NM_001185107.1
NP_001172036.1
4
947
NM_001185108.1
NP_001172037.1
4
782
NM_001185110.1
NP_001172039.1
3
485
NM_001185112.1
NP_001172041.1
3
650
NM_001185113.1
NP_001172042.1
3
485
NM_001185114.1
NP_001172043.1
4
920
NM_001185115.1
NP_001172044.1
4
947
NM_030893.3
NP_112155.2
5
1217
XM_011510133.2
XP_011508435.1
5
1591
XM_011510134.2
XP_011508436.1
5
1652
GenBank mRNA
AJ289111.1
-
5
924
AJ289112.1
-
5
924
AJ289113.1
-
5
820
AJ289114.1
-
5
759
AJ289115.1
-
4
654
AJ289116.1
-
4
654
AJ289117.1
-
4
489
AJ289118.1
-
3
357
AJ289119.1
-
3
357
AJ289120.1
-
3
253
AJ289121.1
-
3
192
AJ289122.1
-
3
192
AK303088.1
-
4
1321
AK303108.1
-
5
1019
AK303233.1
-
2
849
AK311643.1
-
4
836
BC131693.1
-
5
928
40120328
X79517.1
-
2
60
X79519.1
-
1
16
Back To Top
More Information
Panther Classification:
Molecular Function -
major histocompatibility complex protein
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
immune response
antigen processing and presentation, exogenous lipid antigen via MHC class Ib
beta-2-microglobulin binding
endogenous lipid antigen binding
exogenous lipid antigen binding
lipopeptide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0