Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERBB2 SIRNA Assays ›
          Gene Symbol:
          Erbb2
          Gene Name:
          v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian)
          Gene Aliases:
          Erbb-2, HER-2, HER2, Neu, c-erbB2, c-neu, mKIAA3023
          Chromosome Location:
          Chr.11: 98412484 - 98437716 on Build GRCm38
          UniGene ID:
          Mm.290822
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS203967
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Erbb2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003817.1 NP_001003817.1 17 2169
          GenBank mRNA AK031099.1 - 17 2037
          AK031542.1 - 9 867
          AK083669.1 - 17 2038
          AK129487.1 - 17 2139
          AK142296.1 - 17 2163
          BC046553.1 - 3 524
          BC046811.1 - 17 2160 5702040
          BC053078.1 - 17 2160 5702040

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          signal transduction
          cell surface receptor signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          nervous system development
          peripheral nervous system development
          heart development
          neuromuscular junction development
          motor neuron axon guidance
          positive regulation of cell proliferation
          glial cell differentiation
          phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          phosphorylation
          peptidyl-tyrosine phosphorylation
          positive regulation of cell growth
          regulation of microtubule-based process
          negative regulation of immature T cell proliferation in thymus
          wound healing
          myelination
          negative regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          estrous cycle
          positive regulation of translation
          positive regulation of cell adhesion
          positive regulation of transcription from RNA polymerase I promoter
          positive regulation of transcription from RNA polymerase III promoter
          positive regulation of Ras protein signal transduction
          protein autophosphorylation
          sympathetic nervous system development
          response to axon injury
          oligodendrocyte differentiation
          positive regulation of epithelial cell proliferation
          regulation of ERK1 and ERK2 cascade
          nucleotide binding
          RNA polymerase I core binding
          glycoprotein binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          transmembrane signaling receptor activity
          protein binding
          ATP binding
          protein C-terminus binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          growth factor binding
          protein phosphatase binding
          ubiquitin protein ligase binding
          identical protein binding
          protein heterodimerization activity
          Hsp90 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline