Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025602.3 | NP_001020773.1 | 3 | 607 | ||
NM_001294171.1 | NP_001281100.1 | 3 | 440 | |||
NM_010743.3 | NP_034873.2 | 3 | 607 | |||
GenBank mRNA | AK033417.1 | - | 3 | 460 | ||
AK145118.1 | - | 3 | 461 | |||
BC051445.1 | - | 3 | 417 | 30073421 | ||
D13695.1 | - | 2 | 299 | |||
M24843.1 | - | 3 | 448 | |||
Y07519.1 | - | 3 | 451 |