Search
Search

Dhrs4| Gene Symbol: Dhrs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037938.2 | NP_001033027.2 | 3 | 518 | ||
| NM_030686.2 | NP_109611.1 | 3 | 518 | |||
| GenBank mRNA | AB045132.1 | - | 3 | 430 | ||
| AK018788.1 | - | 3 | 401 | |||
| AK146988.1 | - | 3 | 398 | |||
| BC003484.1 | - | 3 | 353 | 2811468 | ||
| BC054361.1 | - | 3 | 479 | 5008299 | ||