Search
Search

Reg3a| Gene Symbol: Reg3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011259.1 | NP_035389.1 | 4 | 360 | ||
| GenBank mRNA | AK008050.1 | - | 1 | 15 | ||
| AK008140.1 | - | 1 | 15 | |||
| AK008470.1 | - | 2 | 195 | |||
| BC119153.1 | - | 4 | 360 | 8733902 | ||
| D63356.1 | - | 4 | 360 | |||
| D63357.1 | - | 4 | 309 | |||