Search
Search

Gngt1| Gene Symbol: Gngt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010314.2 | NP_034444.1 | 2 | 139 | ||
| GenBank mRNA | AK020863.1 | - | 2 | 141 | ||
| AK020903.1 | - | 2 | 140 | |||
| BC023064.1 | - | 2 | 129 | |||
| BC025929.1 | - | 2 | 334 | 4511672 | ||
| U38495.1 | - | 1 | 36 | |||