Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031525.1 | NP_113713.1 | 19 | 2990 | ||
XM_006254789.2 | XP_006254851.1 | 19 | 3080 | |||
XM_008772084.1 | XP_008770306.1 | 18 | 2631 | |||
GenBank mRNA | AF359356.1 | - | 9 | 1043 | ||
AY090783.1 | - | 19 | 2990 | |||
DQ979389.1 | - | 13 | 1729 |