Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015036.1 | NP_001015036.1 | 4 | 980 | ||
XM_006233982.2 | XP_006234044.1 | 5 | 1419 | |||
XM_006233984.2 | XP_006234046.1 | 6 | 1586 | |||
XM_006233985.2 | XP_006234047.1 | 5 | 1421 | |||
XM_006233987.2 | XP_006234049.1 | 6 | 1051 | |||
XM_006233988.2 | XP_006234050.1 | 6 | 1055 | |||
XM_008761770.1 | XP_008759992.1 | 7 | 1263 | |||
XM_008761771.1 | XP_008759993.1 | 7 | 1266 | |||
GenBank mRNA | AJ646881.1 | - | 5 | 864 | ||
BC091354.1 | - | 4 | 980 | 7317530 |