Search
Search

UGT8| Gene Symbol: UGT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128174.2 | NP_001121646.1 | 6 | 3454 | ||
| NM_001322112.1 | NP_001309041.1 | 7 | 3557 | |||
| NM_001322113.1 | NP_001309042.1 | 6 | 3537 | |||
| NM_001322114.1 | NP_001309043.1 | 6 | 3383 | |||
| NM_003360.4 | NP_003351.2 | 5 | 3393 | |||
| GenBank mRNA | AL137342.1 | - | 1 | 924 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL118812.1 | - | 1 | 924 | Chr. 4 - 114676619 - 114678228 [+] on Build GRCh38 | |