Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CASC7 SIRNA Assays ›
          Gene Symbol:
          CASC7 AGO2
          Gene Name:
          cancer susceptibility candidate 7 (non-protein coding)
          argonaute 2, RISC catalytic component
          Gene Aliases:
          LINC00980
          EIF2C2, Q10
          Chromosome Location:
          -
          Chromosome Location:
          Chr.8: 140520158 - 140642406 on Build GRCh38
          UniGene ID:
          Hs.660189
          Hs.743313
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n366831

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CASC7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK092172.1 - 1 617
          Gene Symbol: AGO2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011516965.2 XP_011515267.1 19 8856
          XM_011516966.2 XP_011515268.1 19 6597
          XM_011516968.2 XP_011515270.2 19 5931
          XM_017013317.1 XP_016868806.1 18 5549

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          translation
          translational initiation
          Wnt signaling pathway, calcium modulating pathway
          post-embryonic development
          RNA secondary structure unwinding
          miRNA metabolic process
          gene silencing by RNA
          pre-miRNA processing
          siRNA loading onto RISC involved in RNA interference
          posttranscriptional gene silencing by RNA
          production of miRNAs involved in gene silencing by miRNA
          miRNA mediated inhibition of translation
          mRNA cleavage involved in gene silencing by miRNA
          miRNA loading onto RISC involved in gene silencing by miRNA
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of translational initiation
          phosphatidylinositol-mediated signaling
          positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
          RNA phosphodiester bond hydrolysis, endonucleolytic
          mRNA cleavage involved in gene silencing by siRNA
          positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          RNA 7-methylguanosine cap binding
          RNA polymerase II core binding
          core promoter binding
          double-stranded RNA binding
          single-stranded RNA binding
          mRNA binding
          translation initiation factor activity
          endoribonuclease activity
          protein binding
          protein C-terminus binding
          siRNA binding
          miRNA binding
          poly(A) RNA binding
          metal ion binding
          endoribonuclease activity, cleaving siRNA-paired mRNA
          endoribonuclease activity, cleaving miRNA-paired mRNA

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0