Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IGHM SIRNA Assays ›
          Gene Symbol:
          IGHM
          Gene Name:
          immunoglobulin heavy constant mu
          Gene Aliases:
          AGM1, MU, VH
          Chromosome Location:
          Chr.14: 105851966 - 105856217 on Build GRCh38
          UniGene ID:
          Hs.510635
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n368803

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IGHM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC001872.1 - 5 1039
          BC002963.1 - 5 1019
          BC006180.1 - 5 1039
          BC009851.2 - 5 1000
          BC011857.2 - 6 1019
          BC015760.1 - 5 1071
          BC017356.2 - 5 1036
          BC019235.2 - 5 1038
          BC020240.1 - 5 1067
          BC073758.1 - 5 1108
          BC073767.1 - 5 1027
          BX161420.1 - 2 1037
          X17115.1 - 6 1111
          X58529.1 - 3 1224
          X67292.1 - 2 580
          X67301.1 - 2 580
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BX408885.2 - 1 227 Chr. 14 - 105854441 - 105855773 [-] on Build GRCh38
          GenBankmRNA BX408883.2 - 1 203 Chr. 14 - 105854441 - 105855714 [-] on Build GRCh38
          GenBankmRNA BX429920.2 - 1 174 Chr. 14 - 105854406 - 105855693 [-] on Build GRCh38
          GenBankmRNA BX450748.1 - 1 233 Chr. 14 - 105854479 - 105855765 [-] on Build GRCh38
          GenBankmRNA CR993111.1 - 1 284 Chr. 14 - 105854531 - 105855820 [-] on Build GRCh38
          GenBankmRNA BX450738.1 - 1 214 Chr. 14 - 105854463 - 105855726 [-] on Build GRCh38
          GenBankmRNA BX429923.2 - 1 209 Chr. 14 - 105854411 - 105855730 [-] on Build GRCh38
          GenBankmRNA BX450750.2 - 1 210 Chr. 14 - 105854460 - 105855739 [-] on Build GRCh38
          GenBankmRNA BQ064827.1 - 1 261 Chr. 14 - 105854539 - 105855809 [-] on Build GRCh38
          GenBankmRNA BX462198.2 - 2 421 Chr. 14 - 105854457 - 105856059 [-] on Build GRCh38
          GenBankmRNA BX408875.1 - 1 216 Chr. 14 - 105854435 - 105855764 [-] on Build GRCh38
          GenBankmRNA BX431790.2 - 1 198 Chr. 14 - 105854474 - 105855732 [-] on Build GRCh38
          GenBankmRNA BQ057939.1 - 1 261 Chr. 14 - 105854420 - 105855809 [-] on Build GRCh38
          GenBankmRNA BX429938.2 - 1 279 Chr. 14 - 105854503 - 105855820 [-] on Build GRCh38
          GenBankmRNA BM691274.1 - 1 45 Chr. 14 - 105854457 - 105855575 [-] on Build GRCh38
          GenBankmRNA BX408884.2 - 1 217 Chr. 14 - 105854457 - 105855739 [-] on Build GRCh38
          GenBankmRNA BX408841.2 - 1 238 Chr. 14 - 105854441 - 105855776 [-] on Build GRCh38
          GenBankmRNA BQ061999.1 - 1 262 Chr. 14 - 105854512 - 105855810 [-] on Build GRCh38
          GenBankmRNA BX429939.2 - 1 238 Chr. 14 - 105854448 - 105855788 [-] on Build GRCh38
          GenBankmRNA BX408855.2 - 1 278 Chr. 14 - 105854494 - 105855823 [-] on Build GRCh38
          GenBankmRNA BX429935.2 - 1 278 Chr. 14 - 105854523 - 105855823 [-] on Build GRCh38
          GenBankmRNA BQ889699.1 - 1 155 Chr. 14 - 105854417 - 105855703 [-] on Build GRCh38
          GenBankmRNA BX429931.2 - 1 216 Chr. 14 - 105854448 - 105855739 [-] on Build GRCh38
          GenBankmRNA BX408878.1 - 1 214 Chr. 14 - 105854457 - 105855726 [-] on Build GRCh38
          GenBankmRNA BQ055589.1 - 1 239 Chr. 14 - 105854532 - 105855787 [-] on Build GRCh38
          GenBankmRNA BC013877.1 - 1909 Chr. - - [-] on Build

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0