Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MICAL1 SIRNA Assays ›
          Gene Symbol:
          Mical1
          Gene Name:
          microtubule associated monooxygenase, calponin and LIM domain containing 1
          Gene Aliases:
          MICAL, MICAL-1, Nical
          Chromosome Location:
          Chr.10: 41475860 - 41487030 on Build GRCm38
          UniGene ID:
          Mm.290431
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s101199

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mical1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164433.1 NP_001157905.1 17 2468
          NM_138315.2 NP_612188.1 18 2687
          GenBank mRNA AK154665.1 - 18 2627
          AK154930.1 - 18 2667
          AK157196.1 - 18 2630
          AK159369.1 - 17 2415
          AK170382.1 - 18 2627
          AK170827.1 - 18 2659
          AK170874.1 - 18 2669
          AK170881.1 - 18 2653
          AK171234.1 - 18 2654
          AK171429.1 - 18 2666
          AK171479.1 - 18 2660
          AK171779.1 - 15 3625
          AK172562.1 - 18 2662
          AK172716.1 - 18 2613
          BC021477.1 - 18 2635 5343552
          BC034682.1 - 18 2579 3485807

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          cytoskeleton organization
          sulfur oxidation
          actin filament depolymerization
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          oxidation-reduction process
          actin binding
          monooxygenase activity
          protein binding
          zinc ion binding
          3-(3-hydroxyphenyl)propionate hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
          SH3 domain binding
          Rab GTPase binding
          4-chlorobenzaldehyde oxidase activity
          3,5-xylenol methylhydroxylase activity
          phenylacetate hydroxylase activity
          4-nitrophenol 4-monooxygenase activity
          dimethyl sulfide monooxygenase activity
          alpha-pinene monooxygenase [NADH] activity
          phenanthrene 9,10-monooxygenase activity
          1-hydroxy-2-naphthoate hydroxylase activity
          toluene 4-monooxygenase activity
          xylene monooxygenase activity
          dibenzothiophene monooxygenase activity
          6-hydroxy-3-methyl-2-oxo-1,2-dihydroquinoline 6-monooxygenase activity
          chlorophenol 4-monooxygenase activity
          carbon disulfide oxygenase activity
          toluene 2-monooxygenase activity
          1-hydroxy-2-oxolimonene 1,2-monooxygenase activity
          phenanthrene 1,2-monooxygenase activity
          tetrahydrofuran hydroxylase activity
          styrene monooxygenase activity
          toluene-4-sulfonate monooxygenase activity
          toluene-sulfonate methyl-monooxygenase activity
          3-methyl-2-oxo-1,2-dihydroquinoline 6-monooxygenase activity
          2-hydroxy-phenylacetate hydroxylase activity
          2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA 1,2-monooxygenase activity
          phenanthrene 3,4-monooxygenase activity
          toluene 3-monooxygenase activity
          4-hydroxyphenylacetate,NADH:oxygen oxidoreductase (3-hydroxylating) activity
          limonene monooxygenase activity
          2-methylnaphthalene hydroxylase activity
          1-methylnaphthalene hydroxylase activity
          bisphenol A hydroxylase A activity
          salicylate 5-hydroxylase activity
          isobutylamine N-hydroxylase activity
          branched-chain dodecylbenzene sulfonate monooxygenase activity
          3-HSA hydroxylase activity
          4-hydroxypyridine-3-hydroxylase activity
          2-octaprenyl-3-methyl-6-methoxy-1,4-benzoquinol hydroxylase activity
          6-hydroxynicotinate 3-monooxygenase activity
          metal ion binding
          tocotrienol omega-hydroxylase activity
          FAD binding
          thalianol hydroxylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0