Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139152.1 | NP_631891.1 | 6 | 1360 | ||
GenBank mRNA | AY057054.1 | - | 6 | 1360 | ||
BC107370.1 | - | 4 | 729 | |||
BC107371.1 | - | 5 | 568 | |||
BC127152.1 | - | 4 | 729 | |||
BC141295.1 | - | 5 | 1326 | 9055907 | ||
BC145621.1 | - | 5 | 1326 | 9054189 |