Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172650.2 | NP_766238.1 | 14 | 1576 | ||
GenBank mRNA | AK034380.1 | - | 14 | 1577 | ||
AK049362.1 | - | 14 | 1574 | |||
AK087865.1 | - | 14 | 1577 | |||
AK139700.1 | - | 14 | 1577 | |||
BC061458.1 | - | 14 | 1575 | |||
BC079581.1 | - | 14 | 1563 | 5702630 |