Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160139.1 | NP_001153611.1 | 13, 14 | 2133 | ||
NM_023872.3 | NP_076361.1 | 12, 13 | 2076 | |||
GenBank mRNA | AF263836.1 | - | 12, 13 | 1551 | ||
AK033079.1 | - | 6, 7 | 634 | |||
AK147264.1 | - | 13, 14 | 2136 | |||
AY679158.1 | - | 12, 13 | 1715 |