Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178309.2 | NP_840094.1 | 17 | 2538 | ||
GenBank mRNA | AK033205.1 | - | 17 | 2509 | ||
AK041841.1 | - | 2 | 111 | |||
AK080771.1 | - | 2 | 165 | |||
AK145709.1 | - | 17 | 2539 | |||
AK169440.1 | - | 17 | 2485 | |||
AK169823.1 | - | 16 | 2275 | |||
BC094252.1 | - | 17 | 2487 | 6401109 |