Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VMN1R63 SIRNA Assays ›
          Gene Symbol:
          Vmn1r63 Vmn1r186 Vmn1r187 Vmn1r57 Vmn1r58 Vmn1r60 Vmn1r61 Vmn1r62
          Gene Name:
          vomeronasal 1 receptor 63
          vomeronasal 1 receptor Vmn1r186
          vomeronasal 1 receptor Vmn1r187
          vomeronasal 1 receptor 57
          vomeronasal 1 receptor 58
          vomeronasal 1 receptor 60
          vomeronasal 1 receptor 61
          vomeronasal 1 receptor 62
          Gene Aliases:
          V1R1, V1rd1, V3R1
          -
          -
          EG665150, Gm7519
          V1rd4, V3R4
          EG636697, Gm7184
          Gm7186
          V1rd2, V1rd8, V3R2
          Chromosome Location:
          Chr.7: 5802345 - 5805445 on Build GRCm38
          Chromosome Location:
          Chr.7|NT_166307: 94186 - 95088 on Build GRCm38
          Chromosome Location:
          Chr.7|NT_166307: 148877 - 151977 on Build GRCm38
          Chromosome Location:
          Chr.7: 5220478 - 5221410 on Build GRCm38
          Chromosome Location:
          Chr.7: 5408887 - 5413145 on Build GRCm38
          Chromosome Location:
          Chr.7: 5544197 - 5545099 on Build GRCm38
          Chromosome Location:
          Chr.7: 5610411 - 5611313 on Build GRCm38
          Chromosome Location:
          Chr.7: 5674604 - 5676599 on Build GRCm38
          UniGene ID:
          Mm.160382
          Mm.160381
          Mm.160382
          Mm.480266
          Mm.261621
          Mm.480267
          Mm.479999
          Mm.160381
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s118805

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Vmn1r63
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_030742.1 NP_109667.1 2 1513
          GenBank mRNA AF324866.1 - 2 1513
          BC119575.2 - 1 1342 40058043
          Gene Symbol: Vmn1r186
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167567.1 NP_001161039.1 1 628
          Gene Symbol: Vmn1r187
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167568.1 NP_001161040.1 1 628
          Gene Symbol: Vmn1r57
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166734.1 NP_001160206.1 1 628
          Gene Symbol: Vmn1r58
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_030739.2 NP_109664.2 2 968
          GenBank mRNA AF324869.1 - 2 968
          Gene Symbol: Vmn1r60
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166732.1 NP_001160204.1 1 628
          Gene Symbol: Vmn1r61
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166733.1 NP_001160205.1 1 628
          Gene Symbol: Vmn1r62
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_030741.2 NP_109666.2 1 1346
          GenBank mRNA AF324867.1 - 1 1340
          BC113757.1 - 1 1323 40047296
          BC147681.1 - 1 707 9056654
          BC147684.1 - 1 707 9056657

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0