Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNH1 SIRNA Assays ›
Gene Symbol:
RNH1
Gene Name:
ribonuclease/angiogenin inhibitor 1
Gene Aliases:
RAI, RNH
Chromosome Location:
Chr.11: 494512 - 507283 on Build GRCh38
UniGene ID:
Hs.530687
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12083
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002939.3
NP_002930.2
6
1014
NM_203383.1
NP_976317.1
6
946
NM_203384.1
NP_976318.1
5
773
NM_203385.1
NP_976319.1
5
841
NM_203386.2
NP_976320.1
6
904
NM_203387.2
NP_976321.1
6
972
NM_203388.2
NP_976322.1
5
731
NM_203389.2
NP_976323.1
5
799
XM_011520255.1
XP_011518557.1
6
949
XM_011520257.2
XP_011518559.1
6
1197
XM_011520258.2
XP_011518560.1
6
1165
XM_011520259.2
XP_011518561.1
5
714
XM_011520260.2
XP_011518562.1
6
873
XM_011520261.2
XP_011518563.1
6
947
XM_011520262.2
XP_011518564.1
6
1130
XM_011520263.1
XP_011518565.1
5
681
XM_017018106.1
XP_016873595.1
6
1014
GenBank mRNA
AB209196.1
-
4
2264
AL161967.1
-
4
1949
AL534410.3
-
5
787
AL539148.3
-
5
815
BC000677.2
-
5
701
3349381
BC003506.1
-
5
714
2959514
BC011186.1
-
4
499
BC011500.1
-
5
652
4155804
BC014629.2
-
5
701
3029730
BC024037.1
-
5
714
2959514
BC047730.1
-
6
993
5788220
BQ927358.1
-
5
660
M22414.1
-
5
662
M36717.1
-
6
901
X13973.1
-
6
901
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA catabolic process
negative regulation of catalytic activity
regulation of angiogenesis
protein binding
ribonuclease inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0