Search
Search

ABCE1| Gene Symbol: ABCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040876.1 | NP_001035809.1 | 7 | 1001 | ||
| NM_002940.2 | NP_002931.2 | 7 | 1004 | |||
| GenBank mRNA | AB062293.1 | - | 7 | 678 | ||
| AK315198.1 | - | 7 | 697 | |||
| BC016283.1 | - | 7 | 666 | 3909151 | ||
| BT009779.1 | - | 6 | 561 | |||
| X76388.1 | - | 7 | 678 | |||