Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALB SIRNA Assays ›
          Gene Symbol:
          ALB
          Gene Name:
          albumin
          Gene Aliases:
          ANALBA, FDAH, PRO0883, PRO0903, PRO1341
          Chromosome Location:
          Chr.4: 73404255 - 73421412 on Build GRCh38
          UniGene ID:
          Hs.418167
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1227

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000477.5 NP_000468.1 10 1302
          GenBank mRNA A06977.1 - 10 1304
          AF190168.1 - 10 1229
          AF542069.1 - 10 1262
          AK292755.1 - 10 1267
          AK298437.1 - 10 1295
          AK298461.1 - 10 1012
          AK308044.1 - 10 1145
          AK314794.1 - 10 1270
          AY544124.1 - 6 660
          AY550967.1 - 10 1236
          AY728024.1 - 10 1235
          AY960291.1 - 10 1261
          BC034023.1 - 10 1270 4734617
          BC035969.1 - 6 631 4714468
          BC036003.1 - 10 1270 4724105
          BC041789.1 - 6 694 4766983
          CR749331.1 - 10 1269
          DQ986150.1 - 10 1240
          HQ537426.1 - 9 1157
          V00494.1 - 10 1268
          V00495.1 - 10 1304

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          platelet degranulation
          transport
          receptor-mediated endocytosis
          response to nutrient
          cellular response to starvation
          response to organic substance
          bile acid and bile salt transport
          hemolysis by symbiont of host erythrocytes
          lipoprotein metabolic process
          negative regulation of apoptotic process
          negative regulation of programmed cell death
          sodium-independent organic anion transport
          positive regulation of circadian sleep/wake cycle, non-REM sleep
          response to mercury ion
          maintenance of mitochondrion location
          response to platinum ion
          cellular oxidant detoxification
          DNA binding
          fatty acid binding
          copper ion binding
          protein binding
          drug binding
          zinc ion binding
          toxic substance binding
          antioxidant activity
          oxygen binding
          enzyme binding
          pyridoxal phosphate binding
          identical protein binding
          chaperone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0